Learn More
Fisherbrand™ Safety Coated Clear Boston Round Amber Glass Bottles with White Polypropylene Caps
Type III soda-lime glass is ideal for solvent, chemical or sample storage with plastisol coating for safety are easier to grip and harder to break.
775.00 NOK - 1600.00 NOK
Specifications
Color | Amber |
---|---|
Material | Glass (Type 3) |
Closure Color | White |
Closure Included | Yes |
Mouth | Narrow |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16154497
|
Fisherbrand™
FB02911894 |
120 mL |
1095.00 NOK
Pack of 24 |
Estimated Shipment: 13-01-2026
Log in to see stock. |
|||||
16114467
|
Fisherbrand™
FB02911815 |
240 mL |
775.00 NOK
Pack of 12 |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
16124467
|
Fisherbrand™
FB02911817 |
480 mL |
895.00 NOK
Pack of 12 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||||
16104467
|
Fisherbrand™
FB02911814 |
950 mL |
1600.00 NOK
Pack of 12 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
Description
- Coated in layer of plastisol
- Shatter-resistant coating reduces chance of breakage
- If broken, safety coating will contain contents long enough for disposal
- Non-slip coating also allows greater ease of stability, wet or dry
- Not autoclavable
Specifications
Amber | |
White | |
Narrow | |
PTFE/Polyethylene Foam |
Glass (Type 3) | |
Yes | |
Safety-coated Boston Round Bottle | |
Polypropylene |