Learn More
S100A9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26992-1-AP-20UL
 View more versions of this product
Product Code. 16825315
  • 1696.32 NOK / 20µL
Description
S100A9 (MRP-14) is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis.
Specifications
Specifications
S100A9
Polyclonal
Unconjugated
S100A9
60B8AG, CAGB, Calgranulin B, Calprotectin L1H subunit, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP 14, MRP14, NIF, P14, Protein S100 A9, S100A9
Rabbit
Antigen Affinity Chromatography
RUO
6280
-20°C
Liquid
Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P06702
S100A9
S100A9 Fusion Protein Ag25764