Learn More
RPGRIP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13214-1-AP-20UL
 View more versions of this product
Product Code. 16861324
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
This gene encodes a photoreceptor protein that interacts with retinitis pigmentosa GTPase regulator protein and is a key component of cone and rod photoreceptor cells. Mutations in this gene lead to autosomal recessive congenital blindness.
Specifications
Specifications
RPGRIP1
Polyclonal
Unconjugated
RPGRIP1
CORD9, LCA6, RGI1, RGRIP, RPGR interacting protein 1, RPGRIP, RPGRIP1, RPGRIP1d
Rabbit
Antigen Affinity Chromatography
RUO
57096
-20°C
Liquid
Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96KN7
RPGRIP1
RPGRIP1 Fusion Protein Ag4089