Learn More
Greiner Bio-One Round Bottom Polypropylene Culture Tubes
With high thermal, mechanical and chemical stability.
Brand: Greiner Bio-One 184201
This item is not returnable.
View return policy
Description
- Volume: 12mL
- Graduated
- Dia. x Height: 17 x 77mm
- Available sterile and nonsterile
- Cap is sold separately
Specifications
17 mm | |
Vented Snap Cap | |
Polypropylene | |
Non-sterile | |
Round Bottom |
Culture Tube | |
77 mm | |
Yes | |
12 mL | |
No |