Learn More
ROBO2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21635-1-AP-20UL
 View more versions of this product
Product Code. 16891235
  • 1696.32 NOK / 20µL
Description
The ROBO2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ROBO2. This antibody recognizes human, mouse, rat antigen. The ROBO2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ROBO2
Polyclonal
Unconjugated
20 μL
6092
IgG
Western Blot, ELISA
547 μg/mL
Rabbit
Primary
Human, Rat, Mouse