Learn More
BUCHI FlashPure ID C18 Flash Chromatography Cartridges
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
76.65€ - 325.00€
Specifications
Material | Silica - C18 |
---|---|
Type | Flash Cartridge |
For Use With (Application) | Non-Polar Compounds |
Product Line | FlashPure |
Includes: 1 Piece
Product Code | Brand | Bed Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight (Metric) | Price | Quantity & Availability | |||||
16320286
|
BUCHI
145152990 |
4 g |
76.65€
Each |
In Stock
Log in to see stock. ![]() |
|||||
16390276
|
BUCHI
145152103 |
12 g |
106.00€
Each |
Estimated Shipment: 02-06-2025
Log in to see stock. |
|||||
16370306
|
BUCHI
145171532 |
24 g |
146.64€
Each |
Estimated Shipment: 02-06-2025
Log in to see stock. |
|||||
16300286
|
BUCHI
145152104 |
40 g |
182.00€
Each |
Estimated Shipment: 02-06-2025
Log in to see stock. |
|||||
16310286
|
BUCHI
145152105 |
80 g |
260.00€
Each |
Estimated Shipment: 02-06-2025
Log in to see stock. |
|||||
16330286
|
BUCHI
145152991 |
120 g | N/A | ||||||
Description
- High resolution of separation due to high-quality silica
- Increased fraction purity due to a better separation
- Faster evaporation of smaller fraction volumes
- Reproducible performance from batch to batch
- Increased sample throughput due to high sample loading capacity
- Solvent saving: required purification yield is achieved faster
Specifications
Silica - C18 | |
Flash Cartridge | |
Non-Polar Compounds | |
FlashPure |