Learn More
RETSCH Drying Container
Used for Fluid Bed Dryer TG 200.
12840.00 NOK - 44925.00 NOK
Specifications
Type | Drying Container |
---|---|
For Use With (Equipment) | Fluid Bed Dryer TG 200 |
Product Code | Brand | Capacity (Metric) | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Material | Price | Quantity & Availability | ||||
11732305
|
RETSCH
72.002.0005 |
3 x 0.3 L | Glass |
44925.00 NOK
Each |
Estimated Shipment: 22-12-2025
Log in to see stock. |
||||
10248551
|
RETSCH
72.783.0001 |
6 L | Glass |
18720.00 NOK
Each |
Estimated Shipment: 14-10-2025
Log in to see stock. |
||||
10471223
|
RETSCH
72.783.0002 |
6 L | Stainless Steel |
12840.00 NOK
Each |
Estimated Shipment: 29-10-2025
Log in to see stock. |
||||
Description
- Used for Fluid Bed Dryer TG 200
Specifications
Drying Container | |
Fluid Bed Dryer TG 200 |