Learn More
Epredia™ Reticulin Silver Stain Kit
Product Code. 12583667 Shop All Epredia Products
Click to view available options
Type:
Silver Nitrate Solution (10%)
Unit Size:
125mL
Identify and visualize reticular fibers in tissue sections with the Reticulin Silver Stain, a Gordon and Sweet's modification.
  • Working ammoniacal silver solution selectively impregnates and reacts with reticular fibers through multiple reactions, including oxidation and reduction
  • Nuclear Fast Red Stain Solution provides red background stain
TRUSTED_SUSTAINABILITY
Specifications
Type Silver Nitrate Solution (10%)
Quantity 125mL
compliance-icons compliance-icons
Product Identifier
  • SOLN SILVER NITRATE 10% 125ML
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P391-Collect spillage.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contains : Silver nitrate