Learn More
Trajan Scientific and Medical™ Replacement SilTite™ Mini Union Ferrules 
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
Brand:  Trajan Scientific and Medical™ 073470
1106.70 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12937111
  • 1115.00 NOK / Pack of 10
Only null left
Estimated Shipment: 05-09-2025
Description
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
Specifications
Specifications
SilTite Mini Union Ferrules for 0.1 to 0.25 mm ID Columns
0.1 to 0.25 mm
For 0.1 to 0.25 mm Columns
10/Pk.