Learn More
Thermo Scientific™ Remel™ Gram Iodine
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
Brand: Thermo Scientific™ R40057
This item is not returnable.
View return policy
Description
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
- For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
- Selected Gram stains are CE marked
Specifications
Gram Iodine | |
5/Pk. | |
CE Marked | |
5 x 250 mL |