Learn More
Labcold™ Refrigerator RPFG05043 advanced, underbench, glass door with lock
Brand:  Labcold™ RPFG05043
Product Code. 12948624
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Microprocessor controlled temperature regulation for accuracy and stability
  • Temperature range +2°C to +8°C
  • Fan circulated interior
  • Stainless steel construction
  • Door locks fitted as standard
  • Digital temperature display with audio alarm
  • Heavy duty lockable castors on larger models
  • Optimised air circulation
12mm access port also available as a factory fitted option, please enquire.