Learn More
3M™ Refine 300 Series Anti-Scratch, Anti-Fog Safety Glasses
Click to view available options
Frame Color:
Blue
Bronze
Description
- Available in 2 lens colors
- Slim frame design for younger or female users
- Integrated side shield for added protection
- Soft, padded adjustable nose bridge provides a comfortable fit
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Product Type | Safety glasses; Safety Spectacles |
Frame Color | Bronze |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |