Learn More
Quickfit™ Swan Neck Adapter
Has screwthread joint to accept thermometers and tubes
656.00 NOK - 810.00 NOK
Specifications
For Use With (Application) | Ideal for securing thermometers, gas inlet tubes |
---|---|
Material | Borosilicate Glass |
Product Type | Swan neck adapter |
Thread Size | 13 |
Product Code | Brand | Cone Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Price | Quantity & Availability | |||||
12071603
|
Quickfit
MA6/33 |
24/29 |
810.00 NOK
Each |
Estimated Shipment: 22-10-2025
Log in to see stock. |
|||||
12435306
|
Quickfit
MA6/22 |
19/26 |
656.00 NOK
Each |
Estimated Shipment: 22-10-2025
Log in to see stock. |
|||||
Description
Has screwthread joint to accept thermometers and tubes
- Available in different socket, cone, and thread sizes
Specifications
Ideal for securing thermometers, gas inlet tubes | |
Borosilicate Glass | |
Swan neck adapter | |
13 |