Learn More
Quickfit™ Screwthread Adapter to Fit Cone
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
523.00 NOK - 1065.00 NOK
Specifications
For Use With (Application) | Ideal for securing thermometers, gas inlet tubes |
---|---|
Includes | Plastic cap, Silicone Rubber ring and PTFE washer |
Material | Borosilicate Glass |
Product Type | Cone screw thread adapter |
Product Code | Brand | Thread Size | Cone Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Cone Size | Price | Quantity & Availability | ||||
10821900
|
Quickfit
ST52/13 |
13 | 19/26 |
667.00 NOK
Pack of 2 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||
10840580
|
Quickfit
ST54/24 |
24 | 29/32 |
1065.00 NOK
Pack of 2 |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||
10654969
|
Quickfit
ST52/18 |
18 | 19/26 |
769.00 NOK
Pack of 2 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||
10187988
|
Quickfit
ST51/13 |
13 | 14/23 |
667.00 NOK
Pack of 2 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||
10684009
|
Quickfit
ST53/18 |
18 | 24/29 |
826.00 NOK
Pack of 2 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||
10290219
|
Quickfit
ST51/18 |
18 | 14/23 |
523.00 NOK
Pack of 2 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||
10168138
|
Quickfit
ST53/13 |
13 | 24/29 |
826.00 NOK
Pack of 2 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||
10484520
|
Quickfit
ST53/24 |
24 | 24/29 |
687.00 NOK
Pack of 2 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
10426060
|
Quickfit
ST52/24 |
24 | 19/26 |
667.00 NOK
Pack of 2 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||
10494520
|
Quickfit
ST54/13 |
13 | 29/32 |
943.00 NOK
Pack of 2 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||
10870390
|
Quickfit
ST54/18 |
18 | 29/32 |
943.00 NOK
Pack of 2 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||
Description
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
- With screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer
- Ideal for securing thermometers, gas inlet tubes, etc
Specifications
Ideal for securing thermometers, gas inlet tubes | |
Plastic cap, Silicone Rubber ring and PTFE washer | |
Borosilicate Glass | |
Cone screw thread adapter |