Learn More
Quickfit™ Borosilicate Glass Still Head Adapter
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
430.00 NOK - 2790.00 NOK
Specifications
For Use With (Application) | Connecting flask and condenser |
---|---|
Type | Still Head Adapter |
Material | Borosilicate Glass |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
11972098
|
Quickfit
SH4/1 |
Cone Size: 14/23 (Condenser), 14/23 (flask); Socket Size: 14/23 |
472.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12068483
|
Quickfit
SH4/22 |
Cone Size: 19/26 (Condenser), 19/26 (flask); Socket Size: 14/23 |
544.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332115
|
Quickfit
SH4/2 |
Cone Size: 19/26 (Condenser), 19/26 (flask); Socket Size: 19/26 |
687.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12088483
|
Quickfit
SH4/33 |
Cone Size: 24/29 (Condenser), 24/29 (flask); Socket Size: 14/23 |
430.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12078483
|
Quickfit
SH4/23 |
Cone Size: 19/26 (Condenser), 24/29 (flask); Socket Size: 14/23 |
810.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12098483
|
Quickfit
SH4/44 |
Cone Size: 29/32 (Condenser), 29/32 (flask); Socket Size: 14/23 |
790.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15527970
|
Quickfit
SH4/55 |
Cone size: 34/35 (flask), 34/35 (condenser); Socket Size: 14/23 | N/A | ||||||
Beschreibung
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
- Used in distillation assemblies for connecting flasks to condenser
- Upper socket designed to accept thermometer either via cone/screw thread adapter or thermometer pocket
Spezifikation
Connecting flask and condenser | |
Still Head Adapter | |
Borosilicate Glass |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts