Learn More
Corning™ PYREX™ Media Bottle High-Temperature Caps, Open-Top
With drip-free pouring ring
537.67 NOK - 1107.15 NOK
Specifications
Color Red
Material PBT
No. per Case 10/Cs.
No. per Pack 10/Pk.
Sterility Non-sterile
View More Specs
Products 3
Product Code Brand Thread Size (Metric) Price Quantity & Availability  
Product Code Brand Thread Size (Metric) Price Quantity & Availability  
17233474
View Documents
Corning™
1395-25HTSC
GL 25
537.67 NOK
Pack of 10
Only null left
Estimated Shipment: 20-11-2025
Add to basket
17263474
View Documents
Corning™
1395-32HTSC
GL 32
729.34 NOK
Pack of 10
Only null left
Estimated Shipment: 20-11-2025
Add to basket
11759455
View Documents
Corning™
1395-45HTSC
GL 45
1107.15 NOK
Pack of 10
Only null left
Estimated Shipment: 20-11-2025
Add to basket
Description
Description
These are various sizes and types of screw caps for use with storage bottles.
Specifications
Specifications
Red
10/Cs.
Non-sterile
Open Top Cap
PBT
PBT
10/Pk.
Red
Storage bottle
High Temperature