Learn More
PYREX™ Culture Flask Closure
For use with 1135 culture flasks, 1138/10 Fernbach flasks and 1141 baffled flasks.
511.00 NOK - 1630.00 NOK
Specifications
No. per Pack 10
Autoclavable Autoclavable
View More Specs
Products 2
Product Code Brand Closure Material Price Quantity & Availability  
Product Code Brand Closure Material Price Quantity & Availability  
12524917
View Documents
Pyrex
4540/38P
Polypropylene
511.00 NOK
Pack of 10
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 20-11-2025
Add to basket
12687266
View Documents
Pyrex
4550/38M
Aluminum
1630.00 NOK
Pack of 10
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 06-11-2025
Add to basket
Description
Description
  • Two types of closure - Anodised aluminum - Two-position polypropylene
  • Suitable for autoclaving
Specifications
Specifications
10