Learn More
Cole-Parmer™ PVDF Manifold, 2 port
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
1908.00 NOK - 2340.00 NOK
Specifications
Material | PVDF |
---|---|
Min. Operating Temperature (English) | -65°F |
Min. Operating Temperature (Metric) | -54°C |
Maximum Pressure | 110 psi |
Max. Operating Temperature (English) | 230°F |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11735278
|
Cole-Parmer™
31523-31 |
1/4 in. x 1/8 in. |
2340.00 NOK
Each |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
11745278
|
Cole-Parmer™
31523-33 |
3/8 in. x 1/4 in. | |||||||
11755278
|
Cole-Parmer™
31523-35 |
1/2 in. x 3/8 in. | |||||||
Description
- Constructed of durable PVDF.
- Ideal for applications requiring excellent chemical compatibly.
- Available in a variety of configurations
Specifications
PVDF | |
-54°C | |
230°F | |
Manifold |
-65°F | |
110 psi | |
110°C |