Learn More
Thermo Scientific™ PV-1 Performance Verification Solution
Thermo Scientific™ NanoDrop™ PV-1 Performance Verification Solution is used to help ensure the accuracy and reliability of your NanoDrop spectrophotometer data, helping you save time and reduce errors.
Brand:  Thermo Scientific™ CHEM-PV-1
Encompass_Preferred
Product Code. 15747208
  • 174.15€ / Each
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description
Description
Thermo Scientific™ NanoDrop™ PV-1 Performance Verification Solution is used to help ensure the accuracy and reliability of your NanoDrop spectrophotometer data, helping you save time and reduce errors.
Features include:
  • Solution with certified absorbance values
  • Use to create a record of correct photometric performance of a NanoDrop One spectrophotometer
Specifications
Specifications
NanoDrop PV-1 Performance Verification Solution
NanoDrop One and NanoDrop OneC spectrophotometers
Store at room temperature
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • PV-1 NIST Traceable Calibration Check Fluid, 0,5 mL per vial
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Oxidising liquid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Potassium nitrate, Nicotinic acid
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.