Learn More
Bohlender™ PTFE Sleeves
Brand:  Bohlender™ H933-14
770.55 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10546132
  • 825.00 NOK / Each
Only null left
Estimated Shipment: 07-10-2025
Specifications
PTFE
–200 to 250°C
Joint Sleeve
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bohlender™ PTFE Sleeves > Material: PTFE