Learn More
Cole-Parmer™ PTFE Screen Gaskets
Protect your pump, valves and other components in fluid line
Brand:  Cole-Parmer™ 30538-88
 View more versions of this product
Product Code. 16649295
  • 3040.00 NOK / Each
Only null left
Estimated Shipment: 15-09-2025
Description
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Specifications
Specifications
100
5.08 cm
PTFE
PTFE Gasket