Learn More
Lenz Laborglasinstrumente™ Disposable Ground Joint Sleeves, PTFE
Inserts between standard ground glass joints to eliminate the use of grease. Lenz Laborglasinstrumente™ Disposable Ground Joint Sleeves, PTFE provide outstanding chemical resistance at operating temperatures from -200 to +260°.
Brand:  Lenz Laborglasinstrumente™ 01101519
383.18 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10468253
  • 397.00 NOK / Pack of 10
Only null left
Estimated Shipment: 15-09-2025
Description
  • 50μm thickness
  • Simplifies use of joint clips
  • Fits like a second skin without adding bulk
Specifications
Specifications
PTFE
10Pack
NS 19/26
Ground Joint Sleeve
–200 to 260°C