Learn More
Lenz Laborglasinstrumente™ PTFE Ground Joint Sleeve with Grip Collar
Brand:  Lenz Laborglasinstrumente™ 01101714
215.60 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10894531
  • 305.00 NOK / Each
Only null left
Estimated Shipment: 27-08-2025
Specifications
PTFE
1Pack
NS 14/23
Ground Joint Sleeve
–200 to 260°C