Learn More
PRR13 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14101-1-AP-20UL
This item is not returnable.
View return policy
Description
Negatively regulates TSP1 expression at the level of transcription. This down-regulation was shown to reduce taxane-induced apoptosis.Specifications
PRR13 | |
Polyclonal | |
Unconjugated | |
PRR13 | |
proline rich 13, Proline rich protein 13, PRR13, Taxane resistance protein, TXR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54458 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NZ81 | |
PRR13 | |
PRR13 Fusion Protein Ag5280 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |