missing translation for 'onlineSavingsMsg'
Learn More
Potassium titanium oxide, Thermo Scientific Chemicals Product Code.: 11419756

Potassium titanium oxide, Thermo Scientific Chemicals

Product Code. 11419756
2500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11419756

Brand: Thermo Scientific Alfa Aesar B20263.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-97-6
Molecular Formula K2O3Ti
Molecular Weight (g/mol) 174.06
MDL Number MFCD00058803
InChI Key KEWYKJOOCFOYTD-UHFFFAOYSA-N
Synonym kaliumtitanat, titanate,dipotassium, potassiumtitanatewhisker, 2k.o3ti, titanate tio32-,dipotassium, dipotassium;dioxido oxo titanium
PubChem CID 45052161
SMILES [O--].[O--].[O--].[K+].[K+].[Ti+4]

Specifications

Melting Point 1615°C
Quantity 2500 g
IUPAC Name titanium(4+) dipotassium trioxidandiide
Odor Odorless
Chemical Name or Material Potassium titanium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium titanium oxide, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.