Learn More
BRAND™ POM Joint Clip
Will not scratch glass surfaces. Excellent resilience and stability.
Brand: BRAND™ 55646
7.55 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- POM construction
- Outstanding chemical resistance
- Operating temperature up to 150°C
- Fits NS 29/32
Specifications
Joint Clip | |
POM | |
29/32 | |
Red |