Learn More
Sigma Laborzentrifugen™ Polysulfone Sealing Caps
Provides secure closure for centrifuge tubes and buckets.
Brand: Sigma Laborzentrifugen™ 17170
145.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in both round and rectangular shapes
- For use with a variety of bucket and tube sizes
Specifications
Cap | |
Polysulfone | |
Round Bucket 13450, 13650; Polypropylene Tube 800 mL 13656 | |
Round |