Learn More
Deltalab™ Polypropylene Security Screw Cap Containers
Position the tamper evident ring on Deltalab™ Polypropylene Security Screw Cap Containers after screwing the cap in a normal way to ensure the safety of contents in these ultra clear polypropylene containers.
161.12€
Product Code | Brand | Capacity (Metric) | No. per Case | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | No. per Case | Price | Quantity & Availability | ||||
15270996
|
Deltalab™
409726.G |
150 mL | 400/Cs. |
161.12€
Pack of 350 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Ultra clear polypropylene containers with a specially designed tamper evident ring
- Polyethylene
Specifications
Container | |
Polypropylene | |
Screw Cap |