Learn More
BRAND™ Polypropylene Screw Caps
Shop All Brand Products
Click to view available options
Thread Size (Metric):
GL 25
GL 32
GL 45
GL 54
GL 80
For Use With (Equipment):
For use with laboratory bottles
For use with wide-neck bottles
Description
- Manufactured from polypropylene.
- Replacement and spare screw caps for laboratory bottles available in a variety of sizes.
- Pouring rings sold separately.
Specifications
Specifications
Color | Blue |
Material | Polypropylene |
Quantity | 10Pack |
Thread Size (Metric) | GL 32 |
Certified | Yes |
Product Type | Screw Cap |
For Use With (Equipment) | For use with laboratory bottles |