Learn More
Brand™ Polypropylene Pouring Rings
Manufactured from polypropylene. BRAND™ Polypropylene Pouring Rings are for use with GL45 thread laboratory bottles.
Brand: Brand™ 122664
222.28 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from polypropylene.
- For use with GL45 thread laboratory bottles.
Specifications
Polypropylene | |
GL45 | |
Pouring Ring |
10Pack | |
22 mm | |
Fits GL 45 thread laboratory bottles |