Learn More
Cole-Parmer™ Polypropylene Manifold
Expand your flow and line capacity
984.00 NOK - 1165.00 NOK
Specifications
Material | Polypropylene |
---|---|
Min. Operating Temperature (English) | 32°F |
Min. Operating Temperature (Metric) | 0°C |
Tubing Size | 1/2 in. |
Width (Metric) | 3.81 cm |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11715258
|
Cole-Parmer™
31522-17 |
Polypropylene manifold; 6 outlets; 1/2 in. inlet NPT(F) |
984.00 NOK
Each |
Estimated Shipment: 20-06-2025
Log in to see stock. |
|||||
11775258
|
Cole-Parmer™
31522-29 |
Polypropylene manifold; 10 outlets; 1/2 in. inlet NPT(F) | N/A | ||||||
Description
- Constructed of durable polypropylene.
- Available in a variety of configurations to suit a wide range of applications.
Specifications
Polypropylene | |
0°C | |
3.81 cm | |
150 psi | |
99°C |
32°F | |
1/2 in. | |
3.81 cm | |
210°F |