Learn More
Masterflex™ Polyethylene Straight Connector
Designed for quick and easy assembly without tools
280.00 NOK - 348.00 NOK
Specifications
Description | Barb Connection |
---|---|
Autoclavable | Not Autoclavable |
Material | HDPE |
Sterilization Method | Ethylene Oxide |
Color | Translucent |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11765559
|
Masterflex™
30621-79 |
5/8 in. |
280.00 NOK
Pack of 10 |
Estimated Shipment: 26-05-2025
Log in to see stock. |
|||||
11775559
|
Masterflex™
30621-81 |
3/4 in. |
324.00 NOK
Pack of 10 |
Estimated Shipment: 26-05-2025
Log in to see stock. |
|||||
11770049
|
Masterflex™
30621-83 |
1 in. |
348.00 NOK
Pack of 10 |
Estimated Shipment: 26-05-2025
Log in to see stock. |
|||||
Description
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Barb Connection | |
HDPE | |
Translucent |
Not Autoclavable | |
Ethylene Oxide | |
Straight Connector |