Learn More
3M™ Polycarbonate Protective Safety Glasses, 2750 Series
Offers adjustable template length for increased compliance and comfort. The 3M Polycarbonate Protective Safety safety glasses offer excellent impact resistance.
142.38 NOK - 160.00 NOK
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
---|---|
Certifications/Compliance | EN 166:2001 |
Product Type | Safety glasses; Safety Spectacles |
For Use With (Application) | Construction, forestry |
Frame Color | Black |
Description
Provide additional features and style for increased security, comfort, and wearer acceptance.- Excellent impact resistance
- Polycarbonate lens provides reliable protection against specified levels of hazardous UV radiation
- Adjustable temple length to suit the individual
- Anti-scratch and anti-fog coating for increased durability and improved vision
- Ideal for use in many environments including agriculture, assembly work, automotive and bodyshops, construction, DIY, grinding, laboratories, light engineering, and woodworking
Specifications
Anti-fog, Scratch-resistant | |
Safety glasses; Safety Spectacles | |
Black | |
Flat, Adjustable length |
EN 166:2001 | |
Construction, forestry | |
1 Ea. | |
Polycarbonate |