Learn More
PNRC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 51052-1-AP-150UL
 View more versions of this product
Product Code. 16876325
  • 4821.09 NOK / 150µL
Description
Nuclear receptor coactivator. May play a role in signal transduction.
Specifications
Specifications
PNRC1
Polyclonal
Unconjugated
PNRC1
B4 2, PNAS 145, PNRC1, PROL2, Proline rich protein 2, Protein B4 2, PRR2, RP11 63L7.5
Rabbit
Antigen Affinity Chromatography
RUO
10957
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q12796
PNRC1
PNRC1 Fusion Protein Ag0100