Learn More
PNMA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13631-1-AP-150UL
 View more versions of this product
Product Code. 16851944
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
PNMA1 encodes a protein that is highly restricted to the brain and testis. Anti-PNMA1 reacts mainly with subnuclear elements (including the nucleoli) and to a lesser degree the cytoplasm.
Specifications
Specifications
PNMA1
Polyclonal
Unconjugated
PNMA1
37 kDa neuronal protein, MA1, paraneoplastic antigen MA1, PNMA1
Rabbit
Antigen Affinity Chromatography
RUO
170636, 70481, 9240
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8C1C8, Q8ND90, Q8VHZ4
Pnma1
PNMA1 Fusion Protein Ag4549
150 μL
Primary
Human, Mouse, Rat