Learn More
Hecht Karl™ Plastic Rain Gauge
Brand: Hecht Karl™ 44278010
76.25 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
250 mm | |
Holder | |
Clear Plastic | |
Rain Gauge |