Learn More
Sampling Systems™ Plastic Powder Funnels
These Plastic Powder Funnels are ideal for loading powder into drums, containers or kegs.
23.35 NOK - 454.00 NOK
Specifications
Color | White |
---|---|
For Use With (Application) | Loading powder into containers |
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
15899411
|
Sampling Systems™
8023P-100 |
100mm |
23.35 NOK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
|||||
15809421
|
Sampling Systems™
8023P-150 |
150mm |
43.55 NOK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
|||||
15889411
|
Sampling Systems™
8023H-200 |
200mm |
98.20 NOK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
15819421
|
Sampling Systems™
8023P-250 |
250mm |
342.00 NOK
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
15829421
|
Sampling Systems™
8023P-350 |
350mm |
454.00 NOK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||||
Description
- Manufactured from either PP or HDPE
- Crevice-free
- Available in a range of diameter sizes
Specifications
White | |
Loading powder into containers |