Learn More
JP Selecta™ Plactronic Hotplate
The Plactronic rectangular precision hotplate has electronic digital temperature and time control display and conforms to EN 61010, with over-temperature safety cutout.
Brand: JP Selecta™ 6155100
This item is not returnable.
View return policy
Description
- Conforms to EN 61010
- Over-temperature safety cutout
- Digital, push button display
- Pt 100 temperature probe
- Chemically resistant PTFE coated plate
Specifications
PTFE-coated Aluminum Alloy | |
Ambient +5° to 200°C | |
130 mm | |
250 mm | |
Over-temperature Safety Cutout | |
Rectangle | |
EN 61010 | |
420 x 250 x 130 mm |
9.5 x 9.5 in. | |
420 mm | |
7.5 kg | |
700 W | |
Temperature Probe Pt 100 | |
Hotplate | |
Digital |