missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3C2G Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25904-1-AP-20UL
This item is not returnable.
View return policy
Description
Phosphoinositide 3-kinase activity is implicated in assorted cellular responses activated by mammalian cell surface receptors and the regulation of protein sorting in yeast. The p110gamma (PIK3CG) enzyme is activated in vitro by both the alpha and betagamma subunits of heterotrimeric GTP-binding proteins (G proteins) and does not associate with a p85 adaptor molecule. PI 3-kinase C2gamma, also designated p110gamma, may link signaling through G protein-coupled receptors to the generation of phosphoinositide second messengers that are phosphorylated in the D-3 position. The PI 3-kinase C2gamma gene encodes a 1,050 amino acid polypeptide with 36% identity to human PI 3-kinase C2alpha. Research indicates that PI 3-kinase C2gamma can block the growth of human colon cancer cells.Specifications
PIK3C2G | |
Polyclonal | |
Unconjugated | |
PIK3C2G | |
PI3K C2 gamma, PI3K C2GAMMA, PIK3C2G | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5288 | |
-20°C | |
Liquid |
Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75747 | |
PIK3C2G | |
PIK3C2G Fusion Protein Ag23177 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction