Learn More
Thermo Scientific™ Phosphatase Inhibitor Cocktail III
Click to view available options
Quantity:
1 Unit
Unit Size:
1 unit
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Color | White |
Product Type | Phosphatase Inhibitor Cocktail |
Sensitivity | Hygroscopic |
Content And Storage | -30°C to -10°C |
Form | Lyophilized |
Quantity | 1 Unit |