Learn More
Crison™ pH-metro PH 25+
The ideal pH meter for working in the field: lightweight but robust, with IP67 protection.
441.18€ - 464.34€
Specifications
Range (pH) | -2 to 19.99 |
---|---|
Resolution (pH) | 0.01 |
Range (mV) | ±1400 |
Resolution (mV) | 1/0.1 |
Temperature Range (Metric) | -20°C to +150°C |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
12081282
|
Crison™
2500 |
No accessories | N/A | N/A | N/A | ||||
12001292
|
Crison™
2502 |
Buffer solutions, 125mL, pH 4.01, pH 7.00 and pH 9.21 (25°C) Tubes for calibration pH 4.01, pH 7.00 and pH 9.21 |
N/A | N/A | N/A | ||||
Description
- Backlit LCD screen allows simultaneous reading of pH or mV and temperature, also displays the duration of the measurement
- Supplied with a transport case equipped with everything necessary to work both in the field and laboratory
- Designed to hold the calibration jars in a vertical position to facilitate calibration
- Instrument can be in a semi-raised position for better screen viewing
- Calibration bottles allow up to 5 calibrations with only 2mL of buffer
- Intuitive, 6-key keyboard
- Multipin connector for sensors with IP67 sealing
- Auto power off after 5 minutes
- Stability or continuous measurement modes
Specifications
-2 to 19.99 | |
±1400 | |
-20°C to +150°C | |
LCD, Backlit with Pictograms | |
186 x 73 x 38 mm | |
Membrane, 6 keys | |
0°C to 50°C | |
Pt1000, built-in |
0.01 | |
1/0.1 | |
0.1°C | |
67 | |
300 g | |
ABS Enclosure | |
3 x 1.5V AA | |
Over 500 hr. |