Learn More
pH Buffer Solution, Crison Instruments™
19.70€ - 36.25€
Specifications
Boiling Point | Boiling point 100°C |
---|---|
Color | Undesignated |
pH | 9.21 |
Physical Form | Liquid |
Solution Type | Buffer |
Formulation | Borax, also contains germicide |
Odor | Odorless |
Traceability to NIST | Yes |
Solubility Information | In Water at 20°C Complete |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12061462
|
Crison Instruments
LZW9462.99 |
125 mL |
19.70€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12091462
|
Crison Instruments
LZW9465.99 |
250 mL |
36.25€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Boiling point 100°C | |
9.21 | |
Buffer | |
Odorless | |
In Water at 20°C Complete |
Undesignated | |
Liquid | |
Borax, also contains germicide | |
Yes |