Learn More
Petroleum Ether, puriss., Honeywell™
Product Code. p-7073707 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Unit Size:
1 L
2.50 L
25 L
5 L
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Puriss., high boiling, bp 60-80 °C
  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101316-46-5
MDL Number MFCD00081849
Specifications
Density 0.660 to 0.680g/mL at 20°C
Quantity 1 L
Flash Point <−20°C (<−4°F)
UN Number 1268
Grade puriss.
Chemical Name or Material Petroleum Ether