missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD4 Mouse anti-Human, Clone: 1F1A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66100-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein localized to the nucleus in proliferating cells. Expression of this gene is modulated by cytokines in natural killer and T cells. The gene product is thought to play a role in apoptosis but the specific role has not yet been determined. Two transcripts encoding different isoforms have been identified.Specifications
PDCD4 | |
Monoclonal | |
0.74 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q53EL6 | |
PDCD4 | |
PDCD4 Fusion Protein Ag19239 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1F1A11 | |
Unconjugated | |
PDCD4 | |
H731, Nuclear antigen H731 like, PDCD4, Protein 197/15a | |
Mouse | |
Protein G | |
RUO | |
27250 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction