Learn More
PACSIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13219-1-AP-150UL
This item is not returnable.
View return policy
Description
May play a role in vesicle formation and transport.Specifications
PACSIN1 | |
Polyclonal | |
Unconjugated | |
PACSIN1 | |
KIAA1379, PACSIN1, SDPI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23969, 29704, 29993 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BY11 | |
Pacsin1 | |
PACSIN1 Fusion Protein Ag4102 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |