Learn More
Memmert™ Oxygen Control
For Use With: ICO150MED
Brand: Memmert™ ICO150medT6
17085.60 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- N2 connection set includes a hose with coupling and clamp
- For controlling oxygen concentration
- Requires Active Microprocessor Control
Specifications
0.001 | |
ICO150MED | |
Controlling oxygen concentration by N2 inlet |
Adjustment range 1% to 20% O2 | |
Oxygen Control | |
N2 connection set |