Learn More
Thermo Scientific™ Oxoid™ AnaeroJar™ Plate Carrier
The Oxoid AnaeroJar Plate Carrier is for use with AnaeroJar 2.5L, Part No. AG0025A.
Brand: Thermo Scientific™ AG0029A
2050.87 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
OxoidAnaeroJar Plate Carrier is for use with AnaeroJar 2.5L, Part No. AG0025A.
Specifications
AnaeroJar Plate Carrier | |
Thermo Scientific Oxoid | |