Learn More
Grant Instruments™ Optima Bridge Plate
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
Brand:  Grant Instruments™ T-BTS
 View more versions of this product
Product Code. 12797789
  • 2080.00 NOK / Each
Estimated Shipment: 29-07-2025
Explore more special offers
Description
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
  • For use with Grant refrigeration units, heating and cooling circulators
  • Ensures visibility/accessibility of the thermostat whilst optimising bench space
TRUSTED_SUSTAINABILITY
Specifications
Specifications
T Optima Heating Circulator ST5 tank