Learn More
Aqualytic™ Optical Glass Cells
Brand: Aqualytic™ 606890
This item is not returnable.
View return policy
Specifications
Cell | |
Optical Glass | |
Comparator 2000 |
20 mL | |
1 Each |
Product Content Correction
Product Title
Aqualytic™ Optical Glass Cells > Capacity: 20mL