Learn More
Berkshire™ Open-Cell Foam Lab-Tips™ Swabs
General-purpose swab for cleaning small and confined areas
4305.00 NOK - 11175.00 NOK
Product Code | Brand | Length (Metric) | Width (Metric) Tip | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) Tip | Price | Quantity & Availability | ||||
17833232
|
Berkshire™
LTA03163.10 |
76 mm | 5 mm |
7430.00 NOK
Each |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||
17897290
|
Berkshire™
LTA03163.10 |
76 mm | 5 mm |
7430.00 NOK
Pack of 1000 |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||
17863232
|
Berkshire™
LTO70F.20 |
71 mm | 4 mm |
5490.00 NOK
Each |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||
17825510
|
Berkshire™
LTO70P.20 |
71 mm | 3 mm |
5780.00 NOK
Pack of 20 |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||
17873232
|
Berkshire™
LTO70P.20 |
71 mm | 3 mm |
5780.00 NOK
Each |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||
17883232
|
Berkshire™
LTO70R.20 |
71 mm | 3.7 mm |
5515.00 NOK
Each |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||
12876774
![]() |
Berkshire™
LTO70R.20 |
71 mm | 3.7 mm |
6800.00 NOK
Pack of 20 |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||
17853232
|
Berkshire™
LTO1465.10 |
162 mm | 3.8 mm |
4305.00 NOK
Each |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||
12897153
|
Berkshire™
LT000125.10 |
127 mm | 13 mm | ||||||
Description
- Polyurethane foam over rigid core
- 100 ppi open-cell foam
- Thermally bonded head
- Low particles, fibers, ions and extractables, no contaminating adhesives
- Good chemical resistance
- Good sorbency and excellent solvent hold
- Laundered and packaged in cleanroom